Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tissue Adhesive, Permabond 910, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
72590
100491-970EA 59.24 USD InStock
100491-970
Tissue Adhesive, Permabond 910, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Permabond 910 is a tissue adhesive.

  • Contains cyanoacrylate ester
  • Immediate bonding of biological specimens
  • Not water soluble

Permabond 910 is used for mounting specimens for Vibratome sectioning.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty