Would you like to log in again?
Nickel-Plated Forceps with PTFE Coated Tips, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72975 72973
100492-060EA 54.53 USD InStock
100492-060 100492-058
Nickel-Plated Forceps with PTFE Coated Tips, Electron Microscopy Sciences
Forceps
Straight, "PTFE" coated tips, corrosion-free. Offers a soft and good grip to handle delicate articles. Overall length is 98 mm (3 7/8") with medium points.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty