Would you like to log in again?
Curved, PTFE Coated Forceps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72976
100492-062EA 57.85 USD InStock
100492-062
Curved, PTFE Coated Forceps, Electron Microscopy Sciences
Forceps
Full length "PTFE" coated, 98 mm long, serrated, curved tips. No conductivity.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty