Would you like to log in again?
Lab Spoon, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72987
100492-066EA 79.98 USD InStock
100492-066
Lab Spoon, Electron Microscopy Sciences
Spoons
Spoon has 4 mm (⁵/₃₂") Ø stainless steel rod, flattened at one end to give a 8×51 mm blade. The other end has a 13×25 mm spoon.
The blade as well as the spoon is coated with 'PTFE'. Overall length 229 mm (9").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty