Would you like to log in again?
Kodak T-MAX Developer, Liquid, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74245
100492-128EA 40.18 USD InStock
100492-128
Kodak T-MAX Developer, Liquid, Electron Microscopy Sciences
Film Processing Supplies
For use with T-Max Film, Tri-X Pan. A liquid concentrate for easy mixing. Each pack makes 1 gallon.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty