Would you like to log in again?
Graduated Pitchers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74370 74372 74371
100492-170EA 22.48 USD InStock
100492-170 100492-172 100492-174
Graduated Pitchers, Electron Microscopy Sciences
Beakers
Gracefully proportioned, molded pitchers with large comfortable handles and well designed pouring spouts.
  • Metric graduations, and crystal clear to the eye (polymethylpentene)
  • Resists heat to 175°C (374°F)
  • Shatter-proof
  • Excellent chemical resistance
Ideal for preparation of photographic chemicals.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty