Would you like to log in again?
Exhaust Fan, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74850-35 74853 74850-20 74850-40
100494-038EA 798.63 USD InStock
100494-038 100494-036 100494-034 100492-266
Exhaust Fan, Electron Microscopy Sciences
Ventilators
With all of the toxic chemicals and fumes which are released during film processing, the need for adequate ventilation in darkroom work areas cannot be over emphasized.
  • Each fan is one piece molded body and five sections molded louver
  • Different sizes of fresh air supply louvers are available
  • Each size is designed to supply just the right amount of air to a particular model exhaust fan
  • Works best in darkroom that is equipped with fresh air supply louvers or some other kind of air supply
  • Three-wire grounded cord
  • Electrical: 110 V AC, 60 Hz
Improperly ventilated space can be harmful to the health of workers. Now available is our 'Air-No-Lite' 100% light tight darkroom exhaust fans and louvers which insure healthy ventilation continuously. If there is no air supply to the darkroom, the fan will be working against a vacuum and can do nothing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty