Would you like to log in again?
Unique Uni-Band Gripper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
76806
100492-310EA 74.05 USD InStock
100492-310
Unique Uni-Band Gripper, Electron Microscopy Sciences
Forceps
This unique Uni-Band gripper is made of 300 Series Memory stainless steel which is harder than titanium. It is perfect gripper for handling specimen mount stubs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty