Would you like to log in again?
Camera Length, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
80045 80044
100492-366EA 67.65 USD InStock
100492-366 100492-368
Camera Length, Electron Microscopy Sciences
Educational Materials Physics Educational Materials Optics Learning Activities
The nominal value of the effective camera length of an EM perating in the selected area mode is not sufficiently accurate for any calculations of lattice spacing.
The actual value of camera length must be calibrated as the same accelerating voltage and objective lens setting by reference to a known substance with well defined diffraction spacings. The normal specimens are evaporated films on a 3.05 mm grid of aluminum or thallous chloride. The very small crystalline size yields ring patterns suitable for calibration purposes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty