Would you like to log in again?
P47 Scintillators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
82012 82016 82013 82030 82029 82028 82032 82010 82017 82022 82014 82021 82018 82026
100492-396EA 173.94 USD InStock
100492-396 100492-406 100492-408 100492-418 100492-398 100492-420 100492-400 100492-422 100492-424 100492-402 100492-412 100492-404 100492-426 100492-414
P47 Scintillators, Electron Microscopy Sciences
Lights
These scintillators are coated with a thin layer of well selected P47 phosphor (Y1Si2O7:Ce3+; yttrium silicate activated with cerium), and have a high signal output and a good working life. They need not be coated with aluminum prior to use unless cathodoluminescence studies are required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty