Would you like to log in again?
Cassette Racks for Microwave Histoprocessing, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
97050 97051
100492-520EA 695.78 USD InStock
100492-520 100492-522
Cassette Racks for Microwave Histoprocessing, Electron Microscopy Sciences
Wafer Handling Supplies Wafer Separators
Simple and reliable device designed to hold up to 24 standard cassettes for histoprocessing in a microwave. This new rack is manufactured from "PTFE" resins, and will withstand repeated exposure to fixatives, dehydrating agents, clearing agents and paraffin. It is easily cleaned in hot water. It is intended for use with a disposable tray which holds the processing chemicals.
Four processing racks and trays will fit into the EMS 9000 Microwave Processor, so that up to 96 cassettes can be processed in one run. Using this system, the histotechnologist needs to handle the individual cassettes only once, to load them into the racks. The rack is then placed into a tray containing the dehydrating medium, usually 100% ethyl alcohol, and microwaved. After dehydration, the rack containing the cassettes is transferred into a tray containing the clearing agent, usually isopropanol, and microwaved. After clearing, the rack is transferred to a tray containing liquid paraffin, and microwaved. The whole process can take as little as 25 minutes (for small specimens, such as needle biopsies). There are no hazardous chemicals needed (no xylene!).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty