Would you like to log in again?
Square Mesh with Center Reference, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
0250-CU 0100-NI 0075-AU 0050-AU 0075-NI 0200-NI 0400-NI 0050-CU 0300-NI 0200-CU 0200-SS 0400-CU 0300-CU 0100-AU 0150-NI 0400-AU 0100-CU 0150-AU 0150-CU 0075-CU 0250-NI 0200-AU 0050-NI E200-NI 0300-AU
100492-590EA 45.14 USD InStock
100492-590 100492-570 100492-594 100495-546 100492-628 100492-626 100492-604 100492-608 100492-584 100492-620 100492-586 100492-624 100492-602 100492-600 100492-622 100492-582 100492-618 100492-574 100492-630 100492-596 100492-576 100492-578 100492-612 100492-610 100492-632
Square Mesh with Center Reference, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics. Plus, they are the most rigid grids available with a 0.8 mil thickness.
  • 3.05 mm diameter
  • Available in Copper (Cu), Nickel (Ni), and Gold (Au)
  • 50, 75, 100, 150, 200, 250,300, and 400 mesh sizes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty