Would you like to log in again?
Veco Rectangular Slotted Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
1002-CU 0502-NI 1002-NI 0620-CU 0620-NI 0502-CU
100492-636EA 57.73 USD InStock
100492-636 100492-642 100492-656 100492-634 100492-654 100492-644
Veco Rectangular Slotted Grids, Electron Microscopy Sciences
Graticules Reticles and Grids
Veco grids offer superior handling characteristics.
  • Measures 3.05 mm in diameter
Veco grids are the most rigid grids available with a 0.8 mil thickness.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty