Would you like to log in again?
Headband with Magnifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68108-02 68108-15 68108-07 68108-17 68108-20 68108-13 68108-06 68108-03 68108-16 68108-14 68108-05 68108-01 68108-11 68108-04 68108-12
100492-692EA 96.02 USD InStock
100492-692 100492-690 100501-394 100492-704 100492-702 100501-392 100492-708 100492-706 100492-696 100492-694 100492-700 100492-712 100501-396 100492-698 100492-710
Headband with Magnifier, Electron Microscopy Sciences
Magnifiers
Close-up magnification while leaving both hands free for working.
  • Lightweight, adjustable headband for comfort
  • Tilt up and away when not in use
  • Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue
  • Available in a variety of magnifications
The headbands that have two ranges and two magnifications include 3.5X magnification Auxiliary Lens.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty