Would you like to log in again?
Achromatic Magnifier 2 and 4X, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68111-2X 68111-4X
100492-716EA 145.32 USD InStock
100492-716 100492-714
Achromatic Magnifier 2 and 4X, Electron Microscopy Sciences
Magnifiers
The lens diameter is 37 mm and the magnifying power is 4x
The magnifying glass introduced here is the new version of the classic magnifying glass used by the famous detective Sherlock Holmes. This achromatic is more defined in its function and can give you more accurate details of the subject to be observed. This magnifier is composed by two optical glass lenses, which are cemented together. The image seen through this lens is free of color and gives sharp contours so that no vague lines will be found throughout the entire image field. With a solid red wood handle and a red mark at the base of the lens frame, this reminds you to use the face having this red point upwards.
The lens diameter is 37 mm and the magnifying power is 4x. Both faces are purple coated and they measure 39 x 15 x 155 mm. Weight 73g.
Both faces are purple coated and they measure 39 x 15 x 155 mm. Weight 73g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty