Would you like to log in again?
Fluorescent Ruler, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68137-01
100492-722EA 35.63 USD InStock
100492-722
Fluorescent Ruler, Electron Microscopy Sciences
Rulers
It is most useful for all fluorescent viewing and fluorescent photographic work.
Fluorescent Scale in inches and centimeters. This metric/english plastic ruler is printed with a pink phosphorescent ink that glows in the dark.The scale will fluoresce visibly upon illumination with ultraviolet light and serves as a measure of nucleic acid mobility. The scale is 0 to 6" with ¹/₁₆" increments on one edge and 0 to 15 cm with 1 mm increments on the other.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty