Would you like to log in again?
The Economy Rodent Brain Matrix, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
69081-S 69082-S 69080-C 69083-C 69083-S 69080-S 69081-C
100492-858EA 287.89 USD InStock
100492-858 100492-848 100492-856 100492-850 100492-860 100492-854 100492-852
The Economy Rodent Brain Matrix, Electron Microscopy Sciences
Specimens Preserved Specimens Preserved Animal Materials
Low cost acrylic matrices are precisely machined and highly polished to ensure reproducible results. These matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1 mm intervals.
  • Highly polished
  • Precision machined
  • Precise blocking of the brain
  • Reproducible 1 mm segments
  • Cost saving and in all directions
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty