Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Slide Staining Dish for 20 Slides, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

70312-25 70312-22 70312-21 70312-23 70312-24 70312-20
100492-924EA 58.22 USD InStock
100492-924 100492-922 100492-928 100492-926 100492-920 100492-930
Slide Staining Dish for 20 Slides, Electron Microscopy Sciences
Jars Staining Jars
Ideal for immersing slides in several solutions.

The removable glass slide rack has an open bottom which allows for rapid filling and draining. It holds 20 slides-75 x 25 mm (3 x 1"), 75 x 38 mm (3 x 1 1/2"), or 75 x 51 mm (3 x 2") in size. The staining dish measures L x W x H: 107 x 87 x 70 mm. Slide Rack measures: 102 x 80 x 24 mm.

Ordering information: The complete set consists of 1x Staining dish, 1x Cover, 1x Slide Rack, and Handle.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty