Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Staining Dish Set for 30 Slides, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

70312-31 70312-33 70312-30 70312-32
100492-936EA 99.86 USD InStock
100492-936 100492-934 100492-938 100492-932
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Jars Staining Jars
These mix and match staining dish components offer greater flexibility in meeting your requirements. This staining dish accommodates 16, 20 and 30 slide racks.

  • Dish is made from soda-lime glass
  • Holds a variety of sizes: 3 x 1", 3 x 11/2 ", and 3 x 2"
  • Inside: 135 (H) x 102 (W) x 69 (D) mm
  • Outisde: 146 (H) x 113 (W) x 83 (D) mm

Delivery information: Set includes of one staining dish, one lid, one 30-slide stainless steel rack.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty