Would you like to log in again?
Perfect Scale, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70617-10 70616-HD 70617-20
100493-000EA 235.7 USD InStock
100493-000 100493-002 100492-998
Perfect Scale, Electron Microscopy Sciences
Measuring Tapes
The Perfect scale is made from Nickel alloy and 3 mm in length with 1/10 mm divisions. The Scale fits the Perfect Handle (70616-HD).
Delivery information: The Perfect Scale comes complete with one scale and a handle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty