Would you like to log in again?
DuraSeal™ Polyethylene Stretch Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70994-20 70994-14 70994-02 70994-06 70994-01 70994-04 70994-05
100488-114RL 103.27 USD InStock
100488-114 102097-230 103303-504 100501-794 100493-038 102097-226 102097-228
DuraSeal™ Polyethylene Stretch Film, Electron Microscopy Sciences
Seals General Purpose Sealing Films
This Durable heat-resistant, solvent-proof laboratory stretch film forms an airtight heat-resistant barrier.
  • Microwave compatible and tear resistant
  • Chemically resistant to all solvents and caustic agents
  • Withstands a temperature of 100 °C
  • Able to stretch 500 to 800% before breaking or tearing
  • Able to cling to self following stretching
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty