Would you like to log in again?
Nitrogen Purge Cabinet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71219-10 71219-30 71219-20 71219-40
100493-150EA 1448.17 USD InStock
100493-150 100493-152 100501-416 100493-148
Nitrogen Purge Cabinet, Electron Microscopy Sciences
Desiccators Desiccator Cabinets
Acrylic, ethylene propylene, and Type 316 stainless steel welded parts
Simple and easy to use at temperatures up to 120°F. They can be used in freezers, refrigerators, or at room temperature. Cabinets have airtight doors that need no lubrication. Sliding perforated shelves for easy access, transparent acrylic wall for see-through storage. These cabinets are also available with Flow Meters. Direct reading scale flow meters with control levels that let you not only measure flow through the meter, but also make flow adjustments in the line.
Acrylic, ethylene propylene, and Type 316 stainless steel welded parts. With a flow range of 2-20 with an accuracy of ±6%. Flow meter dimensions: 2" scale height x 4.125" overall height with mounting centers of 3".
With a flow range of 2-20 with an accuracy of ±6%. Flow meter dimensions: 2" scale height x 4.125" overall height with mounting centers of 3".
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty