Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Vacuum Pressure Indicator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71242-01
100493-218EA 75.3 USD InStock
100493-218
Vacuum Pressure Indicator, Electron Microscopy Sciences
Vacuum Handling Systems
This Vacuum-Pressure Indicator fits between the desiccator dome and the desiccator stopcock
Its slips easily onto the tapered outlet of the desiccator dome and into the standard desiccator stopcock without requiring any tools.. The thermo-plastic elastometer collapses when there is negative pressure vacuum in the desiccator and expands under ambient atmospheric pressure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty