Would you like to log in again?
Film Casting Device, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71305-01 71305-04 71305-06
100493-270EA 1260.32 USD InStock
100493-270 100493-272 100493-274
Film Casting Device, Electron Microscopy Sciences
Film Processing Supplies
An all glass apparatus that casts uniformly thin films of parlodion, formvar, or butvar directly onto 1 x 3 microscope slides.
  • Film casting solution can be used repeatedly
  • Unit requires 100 mL of film casting solution to start
A built-in fine-pressure-release valve helps control the speed of drainage. The thickness of the film is controlled by the concentration of the film solution and the rate of the drainage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty