Would you like to log in again?
Slide Warmers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71317-10 71317-20
100493-288EA 2432.26 USD InStock
100493-288 102097-466
Slide Warmers, Electron Microscopy Sciences
Slide Warmers/Dryers
These Slide Warmers are designed for cytology, histology, pathology, and other biological applications in industrial and clinical lab environments.
  • Temperature range from slightly above ambient to 65°C
  • Two sizes – Accommodates up to 66 slides
  • Large slide capacity with space-saving design
  • 2 years warranty
  • CE Certified
  • Uniform heat transfer over entire heating surface
  • Black Anodized heavy gauge aluminum worktop for maximum specimen contrast
Model MH6616 temperature ranges slightly above ambient to 100°C, with dry slides across the drying bars, angled on the bar or lying flat without bars
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty