Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

71420-SR 71420-25 71420-DL
100493-306EA 109.52 USD InStock
100493-306 100493-302 100493-304
Glass Staining Dish - 25 Slide Unit, Electron Microscopy Sciences
Jars Staining Jars
These glass staining dishes are made of a heavy glass with snug fit lids.

  • Snug fit lids are included
  • Dishes have gasket seal

The stainless steel rack has a swing handle for easy access and has grooves to take 76 mm microscope slides, up to 40 mm wide horizontally.

Inside Dimensions: 33/4 x 3 x 21/4" (H)
Outside Dimension: 41/4 x 35/8 x 3" (H)

Delivery information: Complete set includes one staining dish with lid and one swing handle rack.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty