Would you like to log in again?
Colored Microscope Slide Boxes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71458-R 71458-B 71459-B 71459-R 71459-W 71458-W
100493-350EA 17.22 USD InStock
100493-350 100493-352 100493-354 100493-344 100493-346 100493-348
Colored Microscope Slide Boxes, Electron Microscopy Sciences
Microscope Slide Boxes
Molded from one piece (lid and bottom) polypropylene these slide boxes are able to accommodate either 25 or 100 standard 75 x 25 mm (3 x 1") slides.
The attached lid is held securely by a closed latch lock. Stacking feet maximize the stability when boxes are stacked up one atop the other. The boxes are unlined and the index number is printed on the bottom. Comes complete with a numbered slide index card. These boxes are available in blue, red or white.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty