Would you like to log in again?
Light Boxes:, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71652 71652-RB 71650 71650-RB
100491-808EA 240.36 USD InStock
100491-808 100491-806 100493-420 100493-422
Light Boxes:, Electron Microscopy Sciences
Lights Light Boxes
Model A: It is compact and can be used on a desk
For viewing and editing slides, transparencies, negatives, micrographs, etc. They also serve as a base for orientation of embedding specimens when observing tissues using a magnifier. Scientifically designed to provide even distribution and uniformity of light. All metal welded construction with a white baked-enamel finish. The viewing surface is a translucent acrylic that is break resistant. It can be used flat on a table, inclined, standing, or hung on the wall. It comes with a ANSI standard 5000K fluorescent lamp, 10 feet cord, on-off switch.
Model A: It is compact and can be used on a desk. Small enough to carry in a briefcase or store in a drawer. Weight 4.5 lbs. Viewing surface measures: 22 x 29.2 cm (8 3/4 x 11 1/2"). Overall measurements: 24.1 x 31.6 x 5 cm (9 1/2 x 12 1/2 x 2").
Model B: It is portable and has a vinyl cover and a heavy duty strap handle. 5000K fluorescent lamp. Weight: 11 lbs.
Viewing surface measures 30.5 x 17.8 cm (12 x 17"). Overall measurements: 36.7 x 45.2 x 8.8 cm (14 1/2 x 18 x 3 1/2").
Model C: The following viewing surfaces are available upon request: 16 x 48" or 24 x 36". Please call for prices. Viewing surface measures: 15"x11½" (38X29.2cm). Overall measurements: 31.6 x 39.2 x 8.8 cm (12 1/4 x 15 1/2 x 3 1/2"). 5000K fluorescent lamp. Weight: 7 lbs.
Model B: It is portable and has a vinyl cover and a heavy duty strap handle. 5000K fluorescent lamp. Weight: 11 lbs.
Viewing surface measures 30.5 x 17.8 cm (12 x 17"). Overall measurements: 36.7 x 45.2 x 8.8 cm (14 1/2 x 18 x 3 1/2").
Model C: The following viewing surfaces are available upon request: 16 x 48" or 24 x 36". Please call for prices. Viewing surface measures: 15"x11½" (38X29.2cm). Overall measurements: 31.6 x 39.2 x 8.8 cm (12 1/4 x 15 1/2 x 3 1/2"). 5000K fluorescent lamp. Weight: 7 lbs.
Small enough to carry in a briefcase or store in a drawer. Weight 4.5 lbs. Viewing surface measures: 22 x 29.2 cm (8 3/4 x 11 1/2"). Overall measurements: 24.1 x 31.6 x 5 cm (9 1/2 x 12 1/2 x 2").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty