Would you like to log in again?
Roto-Shake Genie, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
71834-60 71834-62 71834-51 71834-53 71834-68 71834-10 71834-61 71834-55 71834-57 71834-50 71834-67 71834-66 71834-65 71834-64 71834-52 71834-20 71834-54 71834-56
102097-704EA 29.97 USD InStock
102097-704 100493-448 102097-706 100491-308 100493-462 100493-452 100493-464 100493-454 100493-444 100493-466 100493-456 102097-702 100493-468 100493-446 100493-458 100501-098 100493-460 100493-450
Roto-Shake Genie, Electron Microscopy Sciences
Shakers and Mixers Vortex Mixers
Combination platform/tilt, shaker, rocker, and specialized rotator mixes tubes in 6 different ways: rotating, rocking/shaking, rotating/rocking, rolling/rocking and rotating/rolling, as well as a combination of all these mixing actions
  • Magnetic Platform & Clip Plates
  • Suitable for Use in Cold Rooms & Incubators
  • Change between Rocking & Rotating Quickly & Easily.
  • Economical, Space Saving & Secure
  • 2-Year Warranty
The platform is magnetized to accommodate up to two 15.3 x 15.3 cm (6 x 6") plastic bags per side, a 30.5 x 30.3 cm (12 x 12") non-skid metal rocker tray for conventional rocker/shaker operations, or up to four metal clip plates per side to accommodate similar or different tube sizes for various mixing solutions. Ideal for use in cold rooms and incubators for complete and even soaking of membrane-based blot assays/washings. Unit can be easily cleaned or decontaminated. Maintains the set speed between 32°F (0°C) and 100°F (38°C). Expansion kits are available for increased capacity, and different containers.
The Roto-Shaker Genie is supplied with:
1 non-skid metal rocker tray, measuring 30.5 x 30.5 cm (12 x 12")
4 magnetic stainless steel mounting strips for holding 2 plastic bags, measuring up to 15.3 x 15.3 cm (6 x 6")
1 clip plate for 12 each of 10-13 mm tubes
1 clip plate for 6 each of 15 – 17 mm tubes
1 clip plate for 3 each of 28-30 mm tubes
An optional 3-D Orbital Shaker attachment transforms the unit into an orbital shaker. Easy to remove, two-sided 20.3 x 34.3 cm (8 x 13.5") stainless steel magnetic platform easily converts from rotating to rocking with a simple mechanical adjustment.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty