Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Enviro-Genie® 6-in-1, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71835-10 71835-20 71835-50
100493-470EA 10694.56 USD InStock
100493-470 100493-472 100493-474
Enviro-Genie® 6-in-1, Electron Microscopy Sciences
Shakers and Mixers Shaking Incubators
Applications include the study of microorganisms and tissue cell growth under strictly controlled conditions of temperature and agitation
Designed to provide the security of precise temperature control and uniformity for demanding incubation/refrigeration application without taking up a great deal of space. It uses Thermo-Electric (Peltier) refrigeration and heating with no compressor. Chamber air is gently and continuously recirculated at a rate that ensures the temperature consistency and uniformity of all test samples. Re-circulated air eliminates the possibility of contamination from outside air. Shatter resistant viewing window allows for inspection of samples without effecting the chamber environment.
Applications include the study of microorganisms and tissue cell growth under strictly controlled conditions of temperature and agitation. It performs membrane based assays/washing and hybridizations more efficiently. Microprocessor controls easily achieve desired rotating, rocking, stirring, shaking, temperature and time; all of which are displayed on large LED’s for accurate reproducible results. Timer displays elapsed time or remaining time until programmed off – up to 96 hours. Visible and audible alarms for both temperature and time. Equipped with RS232 port for connection to your computer.
It performs membrane based assays/washing and hybridizations more efficiently. Microprocessor controls easily achieve desired rotating, rocking, stirring, shaking, temperature and time; all of which are displayed on large LED’s for accurate reproducible results. Timer displays elapsed time or remaining time until programmed off – up to 96 hours. Visible and audible alarms for both temperature and time. Equipped with RS232 port for connection to your computer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty