Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Scalpel Handles; Feather®, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72040-04 72040-07 72040-03
100493-604EA 36.53 USD InStock
100493-604 100493-596 100493-598
Scalpel Handles; Feather®, Electron Microscopy Sciences
Scalpels and Scalpel Assemblies Scalpel Handles
Handles for use with different scalpel blades.
  • Handle #3; 100mm long, flat with corrugated grip. For use with scalpel blades #10, #11, #12 and #15.
  • Handle #4; 105mm long, flat with corrugated grip. For use with scalpel blades #20, #21, #22, #23, and #24.
  • Handle #7; 140mm long, slender, contoured with corrugations. For use with scalpel blades #10, #11, #12, and #15
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty