Would you like to log in again?
Coverslip Staining Rack, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72241-01
100493-692EA 124.3 USD InStock
100493-692
Coverslip Staining Rack, Electron Microscopy Sciences
Staining Boxes
The coverslip stainless steel staining rack has grooves to hold 25×25 mm coverslips vertically. It accommodates 25 coverslips at a time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty