Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Corning® Digital Stirring Hotplates, 220 V

Supplier: Electron Microscopy Sciences
Corning®
72350-20 72349-20
100493-720EA 1668.57 USD InStock
100493-720 100493-722
Corning® Digital Stirring Hotplates, 220 V
Stirrers Magnetic Hotplate Stirrers
Corning® digital stirring Hot Plates use innovative microprocessor technology with digital display which optimally controls the heating and stirring to the Pyroceram glass-ceramic stirring hot plates.

  • 220 V model
  • Closed-loop heating and stirring controls that monitor and regulate both temperature and stirring speed
  • Microprocessor maintains consistent and repeatable temperature setting from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
  • Digital LED temperature display is adjustable in 5 °C increments and blinks until temperature is reached
  • Optional external temperature controller truly controls temperature inside the vessel and eliminates the need to closely monitor liquid temperature
  • Exclusive closed-loop stirring control monitors and regulates the stirring speed from 60 to 1150 RPM for aqueous, viscous, or semi-solids solutions
  • Digital LED speed display gives you better control of your stirring process
  • Stronger, larger magnet improves stir bar coupling combined with automatic up speed adjustment minimizes decoupling
  • Stronger, larger magnet improves stir bar coupling combined with automatic up speed adjustment minimizes decoupling

For safety there is a bright LED HOT TOP icon that lights up when top plate is over 60 °C, even when heat control is turned off. Separate temperature sensor provides power cutoff if unit overheats and an optional temperature controller will shut off unit when is not in liquid. A two piece casting design deflects spills away from electronic and includes a built in mount for an optional 18'' (45.7 cm) support rod.

Certifications: UL and cUL approved.

Caution: 220 V model. Corresponding model with voltage suitable for use in US (100493-718) is also available, please see separate product page.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty