Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Pearl® Pipettors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72430-08 72430-04 72430-12 72430-10 72430-14 72430-16 72430-06
100493-778EA 563.04 USD InStock
100493-778 100493-780 100493-782 100493-772 100493-784 100493-774 100493-776
Pearl® Pipettors, Electron Microscopy Sciences
Pipettes Micropipettes
The lightweight, slender design reduces operator fatigue.
  • Entire pipette is autoclavable – no need to recalibrate after autoclaving
  • Highly resistant to chemical reagents
  • Barrel is designed to minimized dead air space for increased pipetter accuracy
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty