Would you like to log in again?
Kendall Filter Pipette Tips (Barrier Tips), Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72460-50 72460-02 72460-30 72460-05 72460-03 72460-60 72460-70 72460-20 72460-10 72460-07 72460-01 72460-04 72460-06 72460-40
100493-800EA 30.11 USD InStock
100493-800 100500-188 100500-178 100493-802 100493-804 100493-806 100500-180 100502-214 100500-182 100500-184 100500-186 100500-176 100502-216 100502-218
Kendall Filter Pipette Tips (Barrier Tips), Electron Microscopy Sciences
Pipette Tips
Kendall filter pipette tips are certified Rnase / Dnase-Free, Human DNA-Free, and PCR inhibitor-free. Their low binding surface reduces DNA and protection retention. Liquid and aerosol barrier prevent pipetter contamination.
  • Types of Membranes
  • Aerosol Barrier: Prevents aerosol contamination
  • Self-sealing Barrier (contains cellulose): Prevents aerosol and liquid contamination
  • Cellulose can contaminate samples
  • Liquid Barrier: Prevents aerosol and liquid contamination
  • Will not contaminate samples
There's a universal fit on most research-grade pipetters. CellPack keeps tips straight for multi-channel pipetters. Made from high-density polyethylene (naturally hydrophobic). Features aerosol, self-sealing and liquid barrier membranes.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty