Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Loctite Adhesive, Electron Microscopy Sciences
Supplier: Henkel
Loctite®
72573-11 72582-01 72570-09 72571-54 72572-61
102098-048EA 15.62 USD InStock
102098-048 100493-808 102098-054 102098-052 102098-050
Loctite Adhesive, Electron Microscopy Sciences
Stains Microscopy and staining reagents
A variety of Loctite adhesives are available that will bond a range of materials.
  • 409™ Super Bonder®
  • 454™ Prism®
  • 861™ Prism®
  • 349™ Impruv®
  • 4011™ Prism®
The 409™ Super Bonder® is a general-purpose gel adhesive, clear, gap fills 0.010”; surface insensitive Ethyl, tensile shear strength 3,200 psi, and temperature range: –65°F to 180°F
The 454™ Prism® is for use with porous surfaces, clear, gap fills 0.010”; surface insensitive Ethyl, gel type, tensile shear strength 3200 psi, temperature range: –65°F to 180°F. Cure speed: fixture: 15 seconds, full: 24 hours.
The 861™ Prism® is for use with flexible surfaces, clear, gap fill 0.008”; surface flexible Alkyl, viscosity 4,000 cP, tensile shear strength 2465 psi, temperature range: –65°F to 212°F. Cure speed: fixture: 25 seconds, full: 24 hours.
The 349™ Impruv® is for bonding glass/metal; appearance: clear/straw, cure type: UV, viscosity: 9,500cP, shore hardness: D70, temperature range: –65°F to 266°F.
Cure speed: fixture: 75 seconds, full: 24 hours.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty