Would you like to log in again?
Dumont Tweezers Style PP, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72878-DS 0508-PP-PO 72878-DB
100493-856EA 79.42 USD InStock
100493-856 102091-764 100493-854
Dumont Tweezers Style PP, Electron Microscopy Sciences
Forceps
Strong tips. Length: 135 mm.
Inox 08 is a medical stainless steel composed of C, Mn, Cr, Mo and V that provides an excellent resistance to corrosion and a good resistance to salt. Although not as hard as Carbon steels, Inox 08 offers excellent resistance to corrosion. This magnetic alloy supports temperatures of around 400°C (DIN 50914) and is suitable for autoclave sterilization at 180°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty