Would you like to log in again?
Magnabrite™ Light-Gathering Magnifiers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Magnabrite™
74393-64 74393-51 74393-90
100494-006EA 152.99 USD InStock
100494-006 100494-004 100494-008
Magnabrite™ Light-Gathering Magnifiers, Electron Microscopy Sciences
Magnifiers
Magnabrite™ magnifies the viewing area by 4X, while intensifying available light and focusing it on the subject area
Made from solid grounded acrylic (not molded) and polished like a glass lens. Crystal clear and pre-focused. There are no bulbs or batteries needed. Available light is nearly four times brighter in the subject area. Available with 90, 64, or 51 mm diameter base.
An "Infinite eyepoint" eliminates the need to keep the eye close to the magnifier. It can be viewed from any distance, at any angle which reduces eye strain and allows several people to view the same object at once. Base is concave which reduces the risk of scratching the film. The light comes with a drawstring storage pouch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty