Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Photographic Darkroom Timer, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74395 74395-22
100502-128EA 500.29 USD InStock
100502-128 100494-010
Photographic Darkroom Timer, Electron Microscopy Sciences
Timers Mechanical Timers
A new improved Model 300 Darkroom Timer shows minutes and seconds in total darkness.
  • Two hands permit quick setting by seconds, minutes, or combinations of seconds and minutes
  • Can be used with a relay for heavier loads
  • Easy to read numerals on a large 6¹/₂" diameter face
  • Luminated by phosphor fluorescence
The 60-minute time range can be set in minutes for developing and seconds for enlarging control. Two separate outlet receptacles control an enlarger or printer and safelight by a combination time/focus switch. Each outlet has a 750-watts capacity.
Input voltage is 100 to 130 VAC, 50 or 60 Hz (U.S. and Canada) and 210 to 250 VAC, 50 or 60 Hz for other countries.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty