Would you like to log in again?
Cross Sectional Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
75942-01
100494-348EA 299.35 USD InStock
100494-348
Cross Sectional Holder, Electron Microscopy Sciences
Specimen Mounting Supplies
Made from non-magnetic stainless steel with 3.1 mm (1/8") diameter pin and adjustable angle turn-screw.
Just insert specimens edge-on and observe the cross section directly.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty