Would you like to log in again?
Lasner-Lamann Tissue Forceps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77893-LM2.5 77893-7LM2.5
100494-430EA 289.91 USD InStock
100494-430 100494-428
Lasner-Lamann Tissue Forceps, Electron Microscopy Sciences
Forceps
Uncompromised slip resistance with automatic tissue manipulation.
  • Ergonomic Featherlite™ spring handle relieves excess pressure and mechanical stress for ideal compression in either hand
  • Patented twisted I-beam design with three point fixation that provides unparralled stability
  • Micro cross-cut carbide jaws provide an extraordinary grip on tissue and suture needles
  • Adjustable auto-lock feature permits auto-retention of tissue
There are two choices in tissue forceps: rat toothed forceps that hold by penetrating a small zone of tissue, or serrated forceps that grasp by inadvertently crushing a broader area and both can lead to microvascular damage. Pressure controlled Lasner-DaMan Tissue Forceps virtually eliminates the penetration or crushing of fragile tissue while providing uncompromised slip resistance.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty