Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77900-S 77901 77902 77900-C
100494-434EA 365.7 USD InStock
100494-434 100492-346 100494-436 100492-348
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Scissors
Featherlite scissors in different styles.
Style DS-Straight Vannas Scissors, 5 mm and smooth blades. Length 125 mm (5").
Style DS-G – Stork Style, 10 mm long and smooth blades. Length: 125 mm (5").
Style DS-GM – Micro Stork Style, 5 mm long and smooth blades. Length: 125 mm (5").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty