Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MicroPoint™ Uniband Scissors and Featherlite Suture Scissors, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77927-7S 77910-11 77910-26 77921-7C 77910-20 77926-6S 77910-36 77921-7S 77910-41 77926-7S 77910-35 77910-16 77926-8C 77910-10 77926-5S 77926-6C 77910-25 77920-5S 77926-8S 77910-1B 77910-30 77910-40 77927-7C 77910-31 77920-5C 77910-15 77926-5C 77915-BS
100494-482EA 303.03 USD InStock
100494-482 100494-460 100494-480 100494-478 100494-456 100494-454 100494-476 100494-452 100494-474 100494-472 100494-450 100494-438 100494-458 100494-492 100494-470 100494-490 100494-446 100494-468 100494-444 100494-488 100494-466 100494-464 100494-486 100494-442 100494-440 100494-484 100494-462 100494-448
MicroPoint™ Uniband Scissors and Featherlite Suture Scissors, Electron Microscopy Sciences
Scissors
A new concept in scissor technology
  • They have three times the shearing angle to reduce tissue crushing
  • They last far longer than ordinary scissors
  • O.R. Castro-Viejo quality at a floor-grade price
  • They offer consistent cutting at the tip
  • The blades cannot work loose
  • The contact between the blades remains constant
  • Ambidextrous: perfect in right and left hands
  • Easy to pickup and use, no clumsy rings
  • Adjustable shearing contact
  • Lifetime Guarantee: All MicroPoint™ Scissors are guaranteed for life against rust, breakage and rivet loosening
These scissors are available in two configurations: Uniband® and FeatherLite®. The MicroPoint Unibands is 5 inches long, one inch wide and half-an-inch tall.
MicroPoint™ scissors can be used for suture or tissue, and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors do not cut by cutting. They cut by shearing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty