Would you like to log in again?
Liston Bone Cutting Forceps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77994-8S 77994-7A 77994-5A 77994-5S 77994-7S 77994-8A
100494-578EA 402.42 USD InStock
100494-578 100494-576 100494-584 100494-574 100494-582 100494-580
Liston Bone Cutting Forceps, Electron Microscopy Sciences
Bone Cutters and Rongeurs
Box joint, available straight or angled. Made from stainless steel with a polished finish.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty