Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Reinforced Plastic Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78190-01 78191-01 78194-01 78193-12 78192-01 78193-01 78191-12 78192-12 78190-12 78194-12
100494-632EA 25.6 USD InStock
100494-632 100499-864 100499-866 100494-640 100499-862 100494-638 100499-870 100499-868 100494-636 100494-634
Reinforced Plastic Tweezers, Electron Microscopy Sciences
Forceps
Reinforced plastic tweezers made of PBTP (Polybutylenterephthalate) with 20% fiber-glass.
  • Good resistance to chemicals and acids
  • Withstand temperature upto 200 °C
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty