Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Wafer Tweezers, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
78225-01 78237-01 78241-01 78239-01 78240-01 78229-01 78220-01 78226-01
100494-654EA 73.54 USD InStock
100494-654 100494-644 100494-652 100494-650 100494-658 100494-648 100494-656 100494-646
Wafer Tweezers, Electron Microscopy Sciences
Forceps
Style 2LB/4: No teeth

Length: 128 mm (5"). B = 12.5 mm (1/2"), C = 8 mm (1/3"), D = 3 mm (1/8"), E = 2 mm (1/12"), F = 4 mm (1/6"), M = 9 mm (1/3"), N = 2 mm (1/12"), R = 48 mm (1 3/4"), S = 42 mm (1 2/3") Made from SA steel with serrated handles. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers. They have a smooth polished non-glare surface with a satin finish. The tweezers have stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping. The tweezers are also available in Peek material. Peek: An ESD Safe, 105Ω x cm. Soft, conductive fiber, with heat resistance up to 250°C (short exposure 300°C).

Style 2LB/4: No teeth. Length: 128 mm (5"). B = 12.5 mm (1/2"), C = 8 mm (1/3"), D = 3 mm (1/8"), E = 2 mm (1/12"), F = 4 mm (1/6"), M = 9 mm (1/3"), N = 2 mm (1/12"), R = 48 mm (1 3/4"), S = 42 mm (1 2/3")

Style 39S-4: 4 teeth, lower part perforated. Length: 128 mm (5"). B =12.5 mm (1/2"), C = 10 mm (1/3"), E = 2 mm (1/12"), F = 6 mm (1/4"), M = 9 mm (1/3"), N = 1.5 mm (1/16"), R = 48 mm (1 3/4"), S = 4 mm (1 2/3")

Style 41LB-2 : 2 teeth. Flat lower paddle. Length: 128 mm (5"). B = 6 mm (1/4"), C = 7.0 mm (1/4"), E = 2 mm (1/12"), F = 3 mm (1/8"), M = 9 mm (1/3"), N = 1.5 mm (1/16"), R = 52 mm (2"), S = 42 mm (1 2/3")

Style 41LB-5: 5 teeth. Flat lower paddle. Length: 132 mm (5 1/4"), B =16 mm (5/8"), C = 9 mm (1/3"), E = 2 mm (1/12"), F = 3 mm (1/8"), M = 11 mm (½"), N = 2 mm (1/12"), R = 52 mm (2"), S = 42 mm (1 2/3")

Style 42LB-6: 6 teeth. Step up lower paddle. Length: 130 mm (5 1/8"), B = 19.5 mm (3/4"), C = 8 mm (1/2"), D = 3 mm (1/8"), E = 3 mm (1/8"), F = 4 mm (1/6"), M = 11 mm (½"), N = 2 mm (1/12"), R = 52 mm (2"), S = 42 mm (1 2/3")

Style 43LB-8-Peek: Clean-room compatible. Length: 138 mm (5 1/2"). B = 27 mm (11/16"), C = 21 mm (3/4"), D = 4 mm (1/6"), E = 6 mm (1/4"), F = 5 mm (1/5"), M = 11 mm (1/2"), N = 1.5 mm (1/16"), R = 61 mm (2 1/3"), S = 35 mm (1 2/3")

Style 43LB-4-Peek: Clean-room compatible. Length: 138 mm (5 1/2"). B = 13 mm (11/16"), C = 21 mm (3/4"), D = 4 mm (1/6"), E = 6 mm (1/4"), F = 5 mm (1/5"), M = 11 mm (1/2"), N = 1.5 mm (1/16"), R = 61 mm (2 1/3"), S = 35 mm (1 3/8")

Style 43LB-10: 10 teeth. Flat lower paddle with opening controller. Length: 190 mm (7 1/2"). B = 33.5 mm (2"), C = 20 mm (3/4"), D = 7 mm (1/4"), E = 2 mm (1/12"), F = 3 mm (1/8"), M = 13 mm (1/2"), N = 2 mm (1/12"), R = 77 mm (3"), S = 42 mm (1 2/3") Style 39S-4: 4 teeth, lower part perforated. Length: 128 mm (5"). B =12.5 mm (1/2"), C = 10 mm (1/3"), E = 2 mm (1/12"), F = 6 mm (1/4"), M = 9 mm (1/3"), N = 1.5 mm (1/16"), R = 48 mm (1 3/4"), S = 4 mm (1 2/3")

Style 41LB-2 : 2 teeth. Flat lower paddle. Length: 128 mm (5"). B = 6 mm (1/4"), C = 7.0 mm (1/4"), E = 2 mm (1/12"), F = 3 mm (1/8"), M = 9 mm (1/3"), N = 1.5 mm (1/16"), R = 52 mm (2"), S = 42 mm (1 2/3")

Style 41LB-5: 5 teeth. Flat lower paddle. Length: 132 mm (5 1/4"), B =16 mm (5/8"), C = 9 mm (1/3"), E = 2 mm (1/12"), F = 3 mm (1/8"), M = 11 mm (½"), N = 2 mm (1/12"), R = 52 mm (2"), S = 42 mm (1 2/3")

Style 42LB-6: 6 teeth. Step up lower paddle. Length: 130 mm (5 1/8"), B = 19.5 mm (3/4"), C = 8 mm (1/2"), D = 3 mm (1/8"), E = 3 mm (1/8"), F = 4 mm (1/6"), M = 11 mm (½"), N = 2 mm (1/12"), R = 52 mm (2"), S = 42 mm (1 2/3")

Style 43LB-8-Peek: Clean-room compatible. Length: 138 mm (5 1/2"). B = 27 mm (11/16"), C = 21 mm (3/4"), D = 4 mm (1/6"), E = 6 mm (1/4"), F = 5 mm (1/5"), M = 11 mm (1/2"), N = 1.5 mm (1/16"), R = 61 mm (2 1/3"), S = 35 mm (1 2/3")
Style 43LB-4-Peek: Clean-room compatible. Length: 138 mm (5 1/2"). B = 13 mm (11/16"), C = 21 mm (3/4"), D = 4 mm (1/6"), E = 6 mm (1/4"), F = 5 mm (1/5"), M = 11 mm (1/2"), N = 1.5 mm (1/16"), R = 61 mm (2 1/3"), S = 35 mm (1 3/8")
Style 43LB-10: 10 teeth. Flat lower paddle with opening controller. Length: 190 mm (7 1/2"). B = 33.5 mm (2"), C = 20 mm (3/4"), D = 7 mm (1/4"), E = 2 mm (1/12"), F = 3 mm (1/8"), M = 13 mm (1/2"), N = 2 mm (1/12"), R = 77 mm (3"), S = 42 mm (1 2/3")

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty