Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EMS High Precisions and Ultra Fine Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78511-27 78314-00D 78317-2AZ 78317-2AX 72919-2AB 78313-00B 78511-0R 72919-2ATA 78517-2 78325-AASA 78320-5TI 78324-10G 78340-51S 78511-0C9 78320-5TE 78360-15A 78320-5X 78322-7 78520-5X 78520-5C 78510-0 78520-5 78512-3C 78319-4 72919-2A 78318-3 78345-57A 78312-000 78511-2AR 78511-01 78512-3 78320-5A 78319-4X 78317-2 78520-5TTH 78318-3CB 78320-5B 78318-3GP 78322-7GP 78320-5C 78325-7R 78320-5AX 78360-15ASW 78319-4A 78512-4R 78322-7E 78325-01 78320-5GP 78319-4R 78320-5 78519-4 78325-5S 78320-5TTH 72919-OA 78325-F 78325-FT 72919-2ATE 78522-7 78520-5A 78316-1 78516-1 78350-62A 78511-1R 78318-3R 78325-5R 78518-3C 78318-3F 78311-00 72919-2ABTE 78318-3C 78325-AC 78322-7TA 78322-7TE 78325-51S 78325-02 78320-5XGP 78518-3 78321-6 78310-0 78328-26 78325-SS6 78325-5BR 78322-7B 78318-3X 78318-3CTA 78325-7A
76376-366EA 68.95 USD InStock
76376-366 76376-244 76376-368 76376-246 76376-202 100494-680 76376-360 100493-872 76376-362 100493-874 76376-240 100493-876 100502-222 100494-710 76375-962 100494-754 100494-752 100494-796 100494-750 101414-022 100494-718 101414-024 100494-716 101414-026 100494-714 100494-758 100501-498 101414-028 100494-712 100494-756 76376-354 76376-356 76376-358 76376-270 76376-232 76376-352 100494-720 100494-764 100494-762 100494-760 101414-030 76499-626 101414-032 100494-728 101414-034 100494-726 101414-036 100494-768 100494-724 101414-038 100494-766 100494-722 102098-998 76376-146 76376-148 100494-818 102098-990 76376-228 100502-886 76376-144 100502-882 76376-220 100494-698 100494-732 101414-040 100502-888 101414-042 100494-770 100494-738 100494-736 100494-734 76376-218 100494-790 100502-890 76376-372 100502-892 76376-210 100494-786 100494-742 100494-820 100494-740 100494-706 100494-704 100494-746 100501-462 100494-744
EMS High Precisions and Ultra Fine Tweezers, Electron Microscopy Sciences
Forceps
Quality tweezers are ideal for laboratory work. For precise handling transmission electron microscopy grid, tweezer styles #3, #3C, #4, #5, #51S, and 5B are recommended.
  • Very popular tweezers
  • Tweezers possess the highest quality
  • These tweezers have an anti-glare satin finish
SA low carbon austenitic steel is non-magnetic and anti-acid. It is ideal for use where corrosion resistance and toughness are primary requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty