Would you like to log in again?
Super Thin and Long Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72919-SS 78353-SSL 78351-65A 78351-66 78351-64A 78348-60
100493-878EA 50.01 USD InStock
100493-878 100494-798 100494-792 100494-804 100494-802 100494-800
Super Thin and Long Tweezers, Electron Microscopy Sciences
Forceps
These long and slender tweezers work well in multiple precision applications and are especially well suited for working near heat sources.
  • Made from SA steel
  • Sharp, relieve tips
  • Double bent tips allow more visibility
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty