Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EMS Wafer Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
4WF.SA.T 78412-39S2 3WL.SA 78414-48WF 78411-38FG 78411-3WF 78414-4WFC 2WFG.SA 78414-43WFG 4WL.SA 5WL.SA 78426-96 78420-8WF 4WFG.SA 2WF.SA 2WL.SA 78414-4WF 45WF.SA.T 78414-44WF 78410-2W 5WF.SA 78425-600M 78411-35S 78414-41W 78410-2WFC 78422-8WNY 78412-39S3
76499-544EA 46.61 USD InStock
76499-544 76499-524 76499-520 76499-542 100494-874 100494-872 100494-850 76499-514 100494-870 100494-858 100494-878 100494-876 100494-854 76499-532 76499-510 76499-512 76499-530 100494-864 100494-862 76499-526 100494-860 100494-882 100494-880 100494-868 101414-016 101414-018 100494-866
EMS Wafer Tweezers, Electron Microscopy Sciences
Forceps
These tweezers are especially designed for the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips etc. Made from antimagnetic/anti acid steel.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty