Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EMS Syntheric Fiber Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78460-702A 78460-710 710.DG 78460-709 78460-708 78460-705 78460-06 78460-707
100494-896EA 13.83 USD InStock
100494-896 100494-886 100494-894 100494-884 100494-892 100494-890 76499-588 100494-888
EMS Syntheric Fiber Tweezers, Electron Microscopy Sciences
Forceps
These fiber tweezers are made from SV plastic which is based on polyvinylidene fluoride (PVDF). Very hard, high tensile and flexural strength. Resistant to high heat and thermal aging. Excellent chemical resistance. ESD safe – low surface resistivity.
  • Good mechanical strength and toughness
  • Smooth surface
  • Continuous use temperature up to 150°C
  • High purity (clean room and medical devices approved, low extraction value)
  • Excellent chemical resistance to most aggressive substances
  • Outstanding resistance to HF, HNO3 , HCl (all concentrations. 90°C)
  • Resistant to UV and radiation (sterilization)
  • ESD safe, very low surface resistively
Typical applications include the handling of very soft and contamination sensitive materials as well as the handling of wafers during chemical processes etc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty